Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111570 Supplier Invitrogen™ Supplier No. PA5111570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVNAGAMSGS GNLMDFLDEP FPDVGTYEDF H Highest antigen sequence indentity to the following orthologs: Rat - 77%, Mouse - 58%.

The family of voltage-dependent chloride channels (CLCs) regulate cellular trafficking of chloride ions, a critical component of all living cells. CLCs regulate excitability in muscle and nerve cells, aid in organic solute transport and maintain cellular volume. The genes encoding human CLC-1 through CLC-7 map to chromosomes 7, 3q26, 4q32, Xp22, Xp11, 1p36 and 16p13, respectively. CLC-1 is highly expressed in skeletal muscle. Mutations in the gene encoding CLC-1 lead to myotonia, an inheritable disorder characterized by muscle stiffness and renal salt wasting. CLC-2 is highly expressed in the epithelia of several organs including lung, which suggests CLC-2 may be a possible therapeutic target for cystic fibrosis. CLC-3 expression is particularly abundant in neuronal tissue, while CLC-4 expression is evident in skeletal and cardiac muscle as well as brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCN4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLCN4
Gene Accession No. P51793
Gene Alias chloride channel 4; chloride channel 4-2; chloride channel protein 4; chloride channel, voltage-sensitive 4; chloride transporter ClC-4; chloride voltage-gated channel 4; CLC 4; CLC4; clC-4; Clc4-2; ClC-4A; CLCN4; Clcn4-2; H(+)/Cl(-) exchange transporter 4; MGC163150; putative chloride channel (similar to Mm Clcn4-2); putative chloride channel 4-2
Gene Symbols CLCN4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CLCN4. Recombinant protein control fragment (Product #RP-107268).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1183
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.