Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCN6 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550934

Catalog No. PA550934


Only null left
Add to Cart

Description

Description

CLCN6 Polyclonal Antibody detects endogenous levels of CLCN6 protein.

This gene encodes a member of the voltage-dependent chloride channel protein family. Members of this family can function as either chloride channels or antiporters. This protein is primarily localized to late endosomes and functions as a chloride/proton antiporter. Alternate splicing results in both coding and non-coding variants. Additional alternately spliced variants have been described but their full-length structure is unknown. [provided by RefSeq, Mar 2012].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLCN6
Polyclonal
Unconjugated
CLCN6
AI850629; chloride channel 6; chloride channel protein 6; chloride channel, voltage-sensitive 6; chloride transport protein 6; chloride voltage-gated channel 6; Clc6; CLC-6; ClC-6d; CLCN6; D1-A2; KIAA0046
Rabbit
Antigen affinity chromatography
RUO
1185, 26372, 295586
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O35454, P51797
CLCN6
Synthesized peptide derived from the Internal region of human CLC-6 (aa 611-660).
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.