Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556927 Supplier Invitrogen™ Supplier No. PA556927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGIYDIHVGL RGVPLLEWET EVEMDKLRAS DIMEPNLTYV YPHTRIQSLV SILRTTVHHA FPVVTENRGN EKEFMKGNQL ISNNIKFKKS SILT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

This gene encodes a member of the voltage-dependent chloride channel protein family. Members of this family can function as either chloride channels or antiporters. This protein is primarily localized to late endosomes and functions as a chloride/proton antiporter. Alternate splicing results in both coding and non-coding variants. Additional alternately spliced variants have been described but their full-length structure is unknown. [provided by RefSeq, Mar 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCN6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLCN6
Gene Accession No. P51797
Gene Alias AI850629; chloride channel 6; chloride channel protein 6; chloride channel, voltage-sensitive 6; chloride transport protein 6; chloride voltage-gated channel 6; Clc6; CLC-6; ClC-6d; CLCN6; D1-A2; KIAA0046
Gene Symbols CLCN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLCN6. Recombinant protein control fragment (Product #RP-95552).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1185
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.