Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLCN7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305136100UL
Description
CLCN7 Polyclonal specifically detects CLCN7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CLCN7 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
1186 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
chloride channel 7, Chloride channel protein 7, CLC-7, CLC7FLJ39644, FLJ26686, H(+)/Cl(-) exchange transporter 7, OPTA2FLJ46423, OPTB4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 626-805 of human CLCN7 (NP_001278.1). | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction