Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLCNKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540989 Supplier Invitrogen™ Supplier No. PA540989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MEEFVGLREG SSGNPVTLQE LWGPCPRIRR GIRGGLEWLK QKLFRLGEDW Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCNKB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLCNKB
Gene Accession No. P51801
Gene Alias chloride channel; chloride channel K1-like; putative basolateral cTAL chloride channel ClC-Ka; chloride channel K2; chloride channel Ka; Chloride channel Kb; chloride channel protein ClC-Ka; Chloride channel protein ClC-Kb; chloride channel, kidney, B; chloride channel, voltage-sensitive Ka; chloride channel, voltage-sensitive Kb; chloride voltage-gated channel Kb; clC-K1; Clck2; ClC-K2; ClC-K2L; Clc-Ka; CLCKB; ClC-Kb; CLCNK; Clcnk1l; CLCNKA; Clcnkb; hClC-Kb; LOC100730738; MGC24087; putative basolateral mTAL chloride channel ClC-Ka; RP11-5P18.8
Gene Symbols CLCNKB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CLCNKB (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1188
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.