Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLDN16 Rabbit anti-Canine, Human, Polyclonal, Proteintech
SDP

Catalog No. 501735724 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5724 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5724 Supplier Proteintech Group Inc Supplier No. 276381AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tight junction group of proteins are a cell-to-cell adhesion structure in epithelial cells that constitute the epithelial junctional complex with adherens junctions and desmosomes. Tight junction strands are mainly composed of claudins, occludin, and JAM. Claudin-16 belongs to the Claudin family of protein which is composed of 23 integral membrane proteins. Claudins are responsible for the formation of tight junction strands and are connected with the actin cytoskeleton mediated by ZO-1. Claudin 16, also known as Paracellin-1, is a renal tight junction protein required for paracellular magnesium resorption. It is localized in the intercellular junctions of the epithelial cells of the thick ascending limb of Henle′s loop (a TAL marker) are used to localize this specific structure of nephrons in kidney sections. Mg2+ resorption occurs in TAL, and Claudin-16 is required for this paracellular function. Failure in renal ′handling′ of magnesium can result in hypermagnesemia, whose major direct toxicity implications are cardiovascular as well as renal failure. Recent studies have shown that the lack of Claudin-16 proteins contributes to the dysfunction of paracellular renal transport systems.

Specifications

Antigen CLDN16
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CLDN16
Gene Accession No. Q9Y5I7
Gene Alias claudin 16, CLDN16, HOMG3, Paracellin 1, PCLN 1, PCLN1
Gene Symbols CLDN16
Host Species Rabbit
Immunogen CLDN16 Fusion Protein Ag26531
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10686, 608218
Target Species Canine, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.