Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLDND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591879 Supplier Invitrogen™ Supplier No. PA591879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Mouse kideny, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: GTDFWYEYRS PVQENSSDLN KSIWDEFISD EADEKTYNDA LFRYNGTVGL WRRCITIPKN MHWYSPPERT ESFDVVTKCV SFTLTEQFME KFVDPGNHNS GIDLLRTYLW R.

CLDND1 is widely distributed in the adult CNS with highest expression in the corpus callosum, caudate nucleus, cerebral cortex, medulla, putamen, spinal cord, substantia nigra and subthalamic nucleus. Weak expression has been detected in the adult heart. There are 2 named isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLDND1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLDND1
Gene Accession No. Q9CQX5
Gene Alias 1110019C08Rik; AA407103; AI849195; AW489850; C3orf4; claudin 25; claudin domain containing 1; claudin domain containing 1 protein; claudin domain-containing protein 1; Claudin-25; Cldn25; CLDND1; D11Moh34; GENX-3745; HSPC174; Membrane protein GENX-3745; MGC111162; MGC3316; MGC9861; PSEC0054; UNQ2511/PRO6000
Gene Symbols CLDND1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-138 of human CLDND1 (Q9NY35).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224250, 288182
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.