Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLDND1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987427
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7427 Supplier Bioss Supplier No. BS6683R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLDND1 is widely distributed in the adult CNS with highest expression in the corpus callosum, caudate nucleus, cerebral cortex, medulla, putamen, spinal cord, substantia nigra and subthalamic nucleus. Weak expression has been detected in the adult heart. There are 2 named isoforms produced by alternative splicing.

Specifications

Antigen CLDND1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CLDND1
Gene Accession No. Q9NY35
Gene Alias 1110019C08Rik; AA407103; AI849195; AW489850; BOS_692; C3orf4; claudin 25; claudin domain containing 1; claudin domain containing 1 protein; claudin domain containing 1 protein-like; Claudin domain-containing protein 1; Claudin-25; Cldn25; CLDND1; D11Moh34; GENX-3745; HSPC174; LOC100355564; Membrane protein GENX-3745; MGC111162; MGC3316; MGC9861; PSEC0054; UNQ2511/PRO6000
Gene Symbols CLDND1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CLDND1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56650
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.