Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLE7 homolog Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318479H
Description
CLE7 homolog Polyclonal antibody specifically detects CLE7 homolog in Human, Mouse samples. It is validated for Western BlotSpecifications
CLE7 homolog | |
Polyclonal | |
Western Blot | |
CGI99, CGI-99, CLE, LCRP369, RLLM1 | |
Recognizes region between residues Between between 75 and 125.. | |
0.1 mL | |
Cancer, Neuroscience | |
51637 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction