Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLE7 homolog Antibody (OTI2A4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271940
Description
CLE7 homolog Monoclonal specifically detects CLE7 homolog in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CLE7 homolog | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
CGI99, CGI-99, CLE, LCRP369, RLLM1 | |
Mouse | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI2A4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
RTRAF | |
Full length human recombinant protein of human C14orf166 (NP_057123) produced in E.coli. | |
100 μg | |
51637 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction