Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Clean-Walk Mat
Shop All 3M Products
Click to view available options
Color:
White
Dimensions (L x W):
36 x 18 in.
45 x 25 in.
46 x 36 in.
Description
- Thin floor mat profile minimizes tripping hazard. Bottom layer of the pad firmly attaches to most smooth, clean surfaces, such as tile or sealed concrete.
- Sticky mat captures dirt on contact before tracking it from one area to another. It prolongs life and appearance of floors and carpet and reduces facility maintenance and clean up. Pad length is sufficient for two step contact.
- Multiple adhesive layers allow for easily renewed surface. Individually numbered tabs ensure that only one layer is removed at a time.
- Facilities, general industrial, maintenance, military, aerospace, construction, transportation, truck, bus, commercial vehicle, manufacturing, automotive, RV, ship yards, metal working, appliance, electronics, food and beverage.
- Protecting, preventing, prolonging life, controlling contamination, reducing dirt and dust, keeping clean, removing dirt

Specifications
Specifications
Dimensions (L x W) | 45 x 25 in. |
Type | Entrance Mat |
Thickness (English) | 0.097 in. |
Color | White |
Material | Acrylic, Polyethylene |
Quantity | 4/Cs. |
Length (English) | 45 in. |
Width (Metric) | 635 mm |
Length (Metric) | 1143 mm |
Width (English) | 25 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction