Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Clean-Wipes™ Cellulose and Polyester Cloth PROMO
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
22.8 x 15.2 cm (9 x 6 in.)
Description
- Reclosable container dispenses single wipes
- Eliminates contamination from dust and dirt
- Premoistened with a blend of pure reagent-grade 70% isopropyl alcohol and 30% deionized water
Specifications
Specifications
| Product Type | IPA Wipes |
| Length (English) | 9 in. |
| Length (Metric) | 22.8 cm |
| Width (English) | 6 in. |
| Width (Metric) | 15.2 cm |
| Material | Polyester Cellulose |
| Dimensions (L x W) | 22.8 x 15.2 cm (9 x 6 in.) |
| For Use With (Application) | Cleaning Optical Parts, Glassware, Electrodes, Cuvettes and Microscopes |
| Saturation Solution | 70% Isopropyl Alcohol, 30% Deionized Water |
| Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction