Learn More
Mopec Cleaning and Maintenance Toolkit
SDP
Keep your grossing station like new with Mopec’s cleaning and maintenance toolkit. This toolkit comes complete with all of the items you need to properly care for your Mopec Maestro™ Encore and your Mopec dissection instruments.
Supplier:  Mopec BU00460
 View more versions of this product
Catalog No. 22-038-247
Only null left
Add to Cart
Description
Description
All Toolkits includes:
  • (1) BE045: SaniPath Disinfectant Foaming Spray
  • (1) BE046: EazySoak Instrument Cleaner
  • (1) BE048: ClearSteel Stainless Steel Cleaner and Polish Spray
  • (1) BE042: PathOdorizer Plus, Deodorizer, Cleaner and Degreaser
  • (1) BF045: Maestro Filter for 48 in. Units
We recommend ordering this toolkit every 3 months.
Specifications
Specifications
Cleaning and Maintenance Toolkit
12 in.
12 in.
60 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.