Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURUS™ Cleanroom Clipboard

Catalog No. 19125953
Click to view available options
Quantity:
20/Cs.

PURUS™ Cleanroom Clipboard is ideal for note taking and data recording in cleanrooms.

  • Antistatic
  • Strong surface with rounded edges
  • UV Coating provides chemical resistance
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanroom
  • Stainless steel clip provides a secure hold
  • Manufactured under strict quality control

Specifications

Material Acrylic
Color Clear
Dimensions (L x W x H) 28.5 x 23.5 x 1.25 cm
For Use With (Application) Cleanrooms
Product Type Clipboard
Quantity 20/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.