Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poltex Cleanroom Sign Sleeve

Click to view available options
Mounting Type:
Magnetic
Self-adhesive
Wall Mount
Description
- PETG sign sleeve.
- Suitable for clean room applications.
- Front face lifts off for easy, thorough cleaning.
- Choose from screw mount, magnet mount, or VHB Tape mount.
Sign sleeve, cleanroom, clean room, Poltex
Specifications
Specifications
Material | Polycarbonate |
Size (English) | 8.5 x 11 in. |
Mounting Type | Magnetic |
For Use With (Application) | Clean Room Applications |
Shape | Rectangular |
Languages | Universal |
Product Type | Sign Sleeve |
Color | Clear |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction