Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Cleanroom Sock Recycling Solution
Shop All Fisherbrand Products

Click to view available options
Dimensions (L x W x H):
12 x 11 x 38 in.
12 x 12 x 20 in.
15 x 13 x 38 in.
Capacity (Metric):
47.20 L
82.20 L
121.43 L
Description
- Recycling solution for disposable cleanroom socks
- All brands of disposable cleanroom socks are accepted
- Eco-friendly Boxes: Made from recycled materials and easy to assemble
- Collection boxes include a self-locking mechanisms and prepaid shipping label
- Do not recycle biological materials, infectious waste, bloodborne pathogens, hazardous materials or socks exposed to more than 10% chemical exposure of the surface area
- Available in 3 sizes: Small 12 x 12 x 20 in., Medium 12 x 11 x 38 in., Large 15 x 13 x 38 in.
- 5 boxes per case
- Quarterly report available upon request: Reports to include, but not limited to, materials recycled, amount processed, and materials end-of-life scenarios.
Specifications
Specifications
Capacity (Metric) | 47.20 L |
Capacity (English) | 10.38 gal. |
Dimensions (L x W x H) | 12 x 12 x 20 in. |
Size | Small |
Autoclavable | Not Autoclavable |
For Use With (Equipment) | Cleanroom Socks |
Material | Recycled Materials |
Product Type | Recycling Box |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction