Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Clear Acrylic Overlaminate for BradyJet Labels

Catalog No. p-200009386 Shop All Brady Corporation Products
Click to view available options
Dimensions (LxW):
10.25 x 7.25 in.
2.42 x 3.6 in.
3.2 x 4.58 in.
4.38 x 6.35 in.
6.35 x 8.55 in.
Temperature Range (English):
-20°F to +180°F
-40°F to +140°F

Achieve up to 10 years of outdoor durability through improved UV and abrasion resistance with acrylic overlaminate for BradyJet labels.

  • No need to cut to size
  • Simply peel and apply with these pre-sized rolls ideally sized to their corresponding BradyJet labels
  • Overlaminate is a quarter-inch wider than its corresponding BradyJet label to ensure complete coverage

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Coating None
Color Clear
Finish Gloss
Format Roll
Material Acrylic
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Pipe and Valve Marking, Warehouse Marking
Min. Operating Temperature (Metric) -29°C
Quantity 340 Labels
Size (Metric) 61.47 x 91.44 mm
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -28.9°C to +82.2°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.076 mm
Dimensions (LxW) 2.42 x 3.6 in.
Length (English) 2.42 in.
Length (Metric) 61.47 mm
Width (English) 3.6 in.
Width (Metric) 91.44 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.