Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plas Labs™ Clear Desiccator with Perforated Shelves

For light sensitive materials
$1802.00 - $4580.00
Catalog Number | Mfr. No. | Height (Metric) | Depth (Metric) | Width (Metric) | No. of Shelves | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (Metric) | Depth (Metric) | Width (Metric) | No. of Shelves | Price | Quantity & Availability | ||
11-389-157
![]() |
Plas Labs™
860CG/NS |
30.4 cm | 30.4 cm | 30.4 cm | 2 |
Each for $1,802.00
|
|
||
11-389-160
![]() |
Plas Labs™
861CG/NS |
60.9 cm | 30.4 cm | 30.4 cm | 4 |
Each for $2,880.00
|
|
||
11-389-163
![]() |
Plas Labs™
862CG/NS |
45.7 cm | 45.7 cm | 45.7 cm | 4 |
Each for $3,470.00
|
|
||
11-389-166
![]() |
Plas Labs™
863CG/NS |
60.9 cm | 45.7 cm | 60.9 cm | 4 |
Each for $4,580.00
|
|
||
Description
- Designed to protect, store, or assist in moving critical materials and prevent contamination
- Dust and moisture free enclosures
- Under positive N2 gas flow, prevent moisture from penetrating molecular structure of stored components
Specifications
Clear | |
Perforated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title