Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Clear Gate Valve Lockout

Catalog No. p-200085128
Click to view available options
For Use With (Equipment):
Valve Handle: 1 to 2 in.
Valve Handle: 2.5 to 5 in.

Gate valve lockout devices deny access and cover the valve handle. This device's unique knockout center fits OS and Y or rising stem valves.

Gate valve lockout devices deny access and cover the valve handle. This device's unique knockout center fits OSY or rising stem valves.

  • Device is made of extremely rugged injection-molded polypropylene for superior impact and chemical resistance.
  • Supplied with English, Spanish and French danger labels.
  • Cover the valve handle with the gate valve lockout device and apply a personal padlock and lockout tag to ensure employee safety.
  • Accepts padlock shackles up to 3/8 in. in diameter.
  • Transparent gate valve covers allow visual inspection of the valve position and condition without having to remove the lockout device.

Specifications

Product Type Lockout Device
Color Clear
Material Polycarbonate
Certifications/Compliance OSHA 1910.147
Description Gate Valve Lockout
For Use With (Equipment) Valve Handle: 1 to 2 in.
Includes Attachable Cleat for Circuit Breaker Lockout Device
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.