Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cleavage stimulation factor 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $666.47
Specifications
| Antigen | CSTF2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Cleavage stimulation factor 2 Polyclonal specifically detects Cleavage stimulation factor 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| CSTF2 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| betaCstF-64 variant 2, CF-1 64 kDa subunit, Cleavage stimulation factor 64 kDa subunit, cleavage stimulation factor subunit 2, Cleavage Stimulation Factor, 3 Pre-RNA, Subunit 2, Cleavage Stimulation Factor, 3 Pre-RNA, Subunit 2, 64kD, Cleavage Stimulation Factor, 3 Pre-RNA, Subunit 2, 64kDa, CSTF 64 KDa Subunit, CstF-64 | |
| CSTF2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1478 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GRGGRDPRGIDARGMEARAMEARGLDARGLEARAMEARAMEARAMEARAMEARAMEVRGMEARGMDTRGPVPGPRGPIPSGMQGPSPINMGAVVPQGSRQVPVMQGTGMQGASIQGGSQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title