Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cleaved PARP (Asp214) Mouse anti-Human, Mouse, R718, Clone: F21-852, BD Horizon™
Mouse Monoclonal Antibody
$182.80 - $402.00
Specifications
Antigen | Cleaved PARP (Asp214) |
---|---|
Clone | F21-852 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
Cleaved PARP (Asp214) Monoclonal antibody specifically detects Cleaved PARP (Asp214) in Human, Mouse samples. It is validated for Intracellular staining (Flow Cytometry)Specifications
Cleaved PARP (Asp214) | |
Flow Cytometry | |
R718 | |
Mouse | |
Human, Mouse | |
Asp214 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
F21-852 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human cleaved PARP | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title