Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC-1 Antibody (MM0187-8K42), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212192AF750
Description
CLEC-1 Monoclonal specifically detects CLEC-1 in Human samples. It is validated for Western Blot.Specifications
CLEC-1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CLEC1A | |
Human recombinant CLEC-1 (extracellular domain) | |
Primary | |
Human | |
IgG2 |
Western Blot | |
MM0187-8K42 | |
Western Blot | |
CLEC-1, CLEC1C-type lectin domain family 1 member A, C-type lectin domain family 1, member A, C-type lectin-like receptor 1, C-type lectin-like receptor-1, MGC34328 | |
Mouse | |
0.1 mL | |
51267 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction