Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC-2/CLEC-1B Antibody (AYP1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250166AF750
Description
CLEC-2/CLEC-1B Monoclonal specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CLEC-2/CLEC-1B | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CLEC1B | |
A recombinant extracellular fragment of human CLEC-2/CLEC-1B (aa 68-229), which is the extracellular domain of the protein | |
0.1 mL | |
51266 | |
Human | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
AYP1 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721 | |
Mouse | |
Protein A purified | |
Primary | |
AYP1 can block the interaction of human CLEC-2 with its endogenous ligand Podoplanin. AYP1 does not cross react with mouse CLEC-2 when tested via FACS. AYP1 does not detect CLEC-2 by western blot. For WB we recommend clone AYP2. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction