Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC-2/CLEC-1B Antibody (AYP2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250167AF700
Description
CLEC-2/CLEC-1B Monoclonal specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Western Blot.Specifications
CLEC-2/CLEC-1B | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLEC1B | |
A recombinant extracellular fragment of human CLEC-2/CLEC-1B (aa 68-229), which is the extracellular domain of the protein. | |
0.1 mL | |
Immunology, Innate Immunity, Signal Transduction | |
51266 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
AYP2 | |
Western Blot | |
1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction