Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC-2/CLEC-1B Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10093458
Description
CLEC-2/CLEC-1B Polyclonal specifically detects CLEC-2/CLEC-1B in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
CLEC-2/CLEC-1B | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
1810061I13Rik, CLEC-2, CLEC2B, CLEC2PRO1384, C-type lectin domain family 1 member B, C-type lectin domain family 1, member B, C-type lectin-like receptor 2, C-type lectin-like receptor-2, QDED721 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_001092901.1; NP_057593.3) | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
NP_001092901.1 | |
CLEC1B | |
Peptide with sequence KGHKCSPCDTNWRY corresponding to internal region according to NP_001092901.1, NP_057593.3. | |
0.1 mg | |
Immunology, Innate Immunity, Signal Transduction | |
51266 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction