Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ CLEC-2, Mouse, Unlabeled, Clone: AYP1,

Catalog No. MABF957MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF957MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF957MI Supplier MilliporeSigma™ Supplier No. MABF957
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C-type lectin domain family 1 member B (UniProt Q9P126; also known as C-type lectin-like receptor 2, CLEC-2) is encoded by the CLEC1B (also known as CLEC2, UNQ721/PRO1384) gene (Gene ID 51266) in human. CLEC-2 is a type II single-transmembrane (a.a. 34-54) protein with a large extracellular (a.a. 55-229) region that contains a C-type lectin domain (a.a. 109-217) and a short N-terminal cytoplasmic tail (a.a. 1-33) with a single hem-immunoreceptor tyrosine-based activation motif (hemITAM; a.a. 7-10). CLEC-2 expression is restricted to platelets, where it mediates platelets activation via its hemITAM motif, leading to proteolytic cleavage of two other platelet ITAM receptors, glycoprotein (GP)VI and Fc RIIa, but not of CLEC-2 itself. CLEC-2 functions as the receptor for the type I transmembrane GP podoplanin that is widely expressed outside of the vasculature, including lymphatic endothelial cells, type 1 lung alveolar cells, lymph node stromal cells, the choroid plexus epithelium, inflammatory macrophages, as well as a subset of activated T-helper (Th)17 cells. Patients with rheumatoid arthritis show increased plasma levels of CLEC-2-positive microparticles. These microparticles are derived from activated platelets and are negative for GPVI, while microparticles in healthy donors are predominantly derived from megakaryocytes and are positive for both CLEC-2 and GPVI. Platelet-specific deletion of CLEC-2 or one of its downstream signaling proteins, Syk, SLP-76, or PLC 2, leads to a number of developmental problems in mice, including blood-lymphatic mixing in midgestation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC-2
Applications Flow Cytometry
Classification Monoclonal
Clone AYP1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in PBS without preservatives.
Gene CLEC1B, CLEC2, UNQ721/PRO1384
Gene Accession No. Q9P126
Host Species Mouse
Immunogen His-tagged recombinant human CLEC-2 extracellular fragment.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_057593
Test Specificity Clone AYP1 targets an extracellular epitope present in both spliced isoforms of human CLEC-2/CLEC1B reported by UniProt (Q9P126).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.