Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC-2A (PILAR) Mouse anti-Human, Brilliant Violet 421, Clone: 705518, BD OptiBuild™

Catalog No. BDB748208 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB748208 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB748208 Supplier BD Biosciences Supplier No. 748208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC-2A (PILAR) Monoclonal antibody specifically detects CLEC-2A (PILAR) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC-2A (PILAR)
Applications Flow Cytometry
Classification Monoclonal
Clone 705518
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CLEC-2A (PILAR)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.