Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC10A/CD301 Antibody (744812), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB48881K
Description
CLEC10A/CD301 Monoclonal antibody specifically detects CLEC10A/CD301 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CLEC10A/CD301 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
744812 | |
Flow Cytometry, CyTOF-ready | |
CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL | |
Mouse myeloma cell line NS0-derived recombinant human CLEC10A/CD301, Gln61-His316, Accession # Q8IUN9 | |
0.1 mL | |
Immunology | |
10462 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction