Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech CLEC11A Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P1621MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P1621MG 1 mg
500P16250UG 50 μg
500P162100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P1621MG Supplier PeproTech Supplier No. 500P1621MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: GHGARGAER EWEGGWGGAQ EEEREREALM LKHLQEALGL PAGRGDENPA GTVEGKEDWE MEEDQGEEEE EEATPTPSSG PSPSPTPEDI VTYILGRLAG LDAGLHQLHV RLHALDTRVV ELTQGLRQLR NAAGDTRDAV QALQEAQGRA EREHGRLEGC LKGLRLGHKC FLLSRDFEAQ AAAQARCTAR GGSLAQPADR QQMEALTRYL RAALAPYNWP VWLGVHDRRA EGLYLFENGQ RVSFFAWHRS PRPELGAQPS ASPHPLSPDQ PNGGTLENCV AQASDDGSWW DHDCQRRLYY VCEFPF. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human SCGF-alpha. Anti-Human SCGF-alpha-specific antibody was purified by affinity chromatography employing an immobilized Human SCGF-alpha matrix. Sandwich ELISA: To detect Human SCGF-alpha by sandwich ELISA (using 100 μL/well) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human SCGF-alpha (500-P162BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human SCGF-alpha. Western Blot: To detect Human SCGF-alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human SCGF-alpha is 1.5-3.0 n g/lane, under either reducing or non-reducing conditions

This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC11A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CLEC11A
Gene Accession No. Q9Y240
Gene Alias AW457320; b SCGF; beta SCGF; betaSCGF; bSCGF; Clec11a; CLECSF3; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 3; C-type lectin domain family 11 member A; C-type lectin domain family 11, member A; C-type lectin superfamily member 3; hCG_22930; LSLCL; lymphocyte secreted C-type lectin; lymphocyte secreted long form of C-type lectin; Osteolectin; P47; SCGF; SCGF b; SCGF beta; SCGF β; SCGFb; SCGFbeta; SCGFβ; stem cell growth factor; stem cell growth factor; lymphocyte secreted C-type lectin; β SCGF; βSCGF
Gene Symbols CLEC11A
Host Species Rabbit
Immunogen E.coli-derived, 33.9 kDa Recombinant Human SCGF-alpha.
Purification Method Antigen affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.