Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571556
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571556 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571556 Supplier Invitrogen™ Supplier No. PA571556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLEC12B Polyclonal Antibody for Western Blot, IHC (P)

Cell surface receptor that protects target cells against natural killer cell-mediated lysis. Modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC12B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CLEC12B
Gene Accession No. Q2HXU8
Gene Alias 4933425B16Rik; Clec12b; C-type lectin domain family 12 member B; C-type lectin domain family 12, member B; Macrophage antigen H; RGD1560487; testis secretory sperm-binding protein Li 205a; UNQ5782; UNQ5782/PRO16089
Gene Symbols CLEC12B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 249-276 amino acids from the C-terminal region of human CLEC12B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387837
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.