Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC14A Antibody (036), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290521AF488
Description
CLEC14A Monoclonal specifically detects CLEC14A in Mouse samples. It is validated for ELISA.Specifications
CLEC14A | |
Monoclonal | |
Alexa Fluor 488 | |
C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGFR5, EGFR-5, Epidermal growth factor receptor 5 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CLEC14A (NP_080085.3; Met1-Thr386). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Mouse CLEC3B Mouse GL1 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
36 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
161198 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction