Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC16A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP181805
Description
CLEC16A Polyclonal specifically detects CLEC16A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CLEC16A | |
Polyclonal | |
Western Blot Reported in scientific literature (PMID: 22002632)., Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q2KHT3 | |
CLEC16A | |
This antibody was developed against Recombinant Protein corresponding to amino acids:VPYFSNLVWFIGSHVIELDDCVQTDEEHRNRGKLSDLVAEHLDHLHYLNDILIINCEFLNDVLTDHLLNRLFLPLYVYSLENQDKGGERPKISLPVSLYLLSQVFL | |
0.1 mL | |
Immunology, Innate Immunity | |
23274 | |
Human, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C-type lectin domain family 16, member A, KIAA0350Gop-1, MGC111457, protein CLEC16A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction