Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC16A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557408 Supplier Invitrogen™ Supplier No. PA557408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPYFSNLVWF IGSHVIELDD CVQTDEEHRN RGKLSDLVAE HLDHLHYLND ILIINCEFLN DVLTDHLLNR LFLPLYVYSL ENQDKGGERP KISLPVSLYL LSQVFL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

This gene encodes a member of the C-type lectin domain containing family. Single nucleotide polymorphisms in introns of this gene have been associated with diabetes mellitus, multiple sclerosis and rheumatoid arthritis. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC16A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLEC16A
Gene Accession No. Q2KHT3
Gene Alias 4932416N17Rik; Clec16a; C-type lectin domain family 16 member A; C-type lectin domain family 16, member A; curt; Gop-1; KIAA0350; MGC111457; mKIAA0350; Protein CLEC16A; RGD1305773
Gene Symbols CLEC16A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLEC16A. Recombinant protein control fragment (Product #RP-94583).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.