Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC18A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597594
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597594 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597594 Supplier Invitrogen™ Supplier No. PA597594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLEC18A Polyclonal Antibody for Western Blot, IHC (P), ELISA

Binds polysaccharides in a Ca(2+)-independent manner with a preferentially binding to fucoidan, beta-glucans and galactans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC18A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CLEC18A
Gene Accession No. A5D8T8, Q7TSQ1
Gene Alias CLEC18A; C-type lectin domain family 18 member A; C-type lectin domain family 18, member A; mannose receptor-like 1; mannose receptor-like protein; Mannose receptor-like protein 2; mannose receptor-like protein 3; Mrcl; MRCL1; Mrlp; MRLP2; RGD1559899
Gene Symbols CLEC18A
Host Species Rabbit
Immunogen Recombinant Human C-type lectin domain family 18 member A protein (27-446AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348174, 353287
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.