Learn More
Invitrogen™ CLEC1A Monoclonal Antibody (2F7)

Description
Peptide Sequence: MQAKYSSTRD MLDDDGDTTM SLHSQGSATT RHPEPRRTEH RAPSSTWRPV ALTLLTLCLV LLIGLAALGL LFFQYYQLSN TGQDTISQME ERLGNTSQEL QSLQVQNIKL AGSLQHVAEK LCRELYNKAG AHRCSPRTEQ WKWHGDNCYQ FYKDSKSWED CKYFCLSENS TMLKINKQED LEFAASQSYS EFFYSYWTGL LRPDSGKAWL WMDGTPFTSE LFHIIIDVTS PRSRDCVAIL NGMIFSKDCK ELKRCVCERR AGMVKPESLH VPPETLGEGD.
Specifications
Specifications
| Antigen | CLEC1A |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CLEC1A |
| Gene Accession No. | Q8NC01 |
| Gene Alias | 5930406N14Rik; CLEC1; CLEC-1; CLEC1A; C-type lectin domain family 1 member A; C-type lectin domain family 1, member A; C-type lectin-like receptor 1; C-type lectin-like receptor-1; RGD1566110; UNQ569/PRO1131 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.