Learn More
Invitrogen™ CLEC1A Polyclonal Antibody

Description
The antibody detects endogenous level of total CLEC1A protein.
Specifications
Specifications
| Antigen | CLEC1A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CLEC1A |
| Gene Accession No. | Q8BWY2, Q8NC01 |
| Gene Alias | 5930406N14Rik; CLEC1; CLEC-1; CLEC1A; C-type lectin domain family 1 member A; C-type lectin domain family 1, member A; C-type lectin-like receptor 1; C-type lectin-like receptor-1; RGD1566110; UNQ569/PRO1131 |
| Gene Symbols | CLEC1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.