Learn More
Invitrogen™ CLEC1B Monoclonal Antibody (17D9), FITC

Description
Rat anti Mouse CLEC2 antibody, clone 17D9 recognizes mouse C-type lectin domain family 1 member B, also known as CLEC2 or CLEC1B.
Specifications
Specifications
| Antigen | CLEC1B |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 17D9 |
| Concentration | 100 μg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide |
| Gene | CLEC1B |
| Gene Accession No. | Q9JL99 |
| Gene Alias | 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.