Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597313 Supplier Invitrogen™ Supplier No. PA597313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVMQRNYLQG ENENRTGTLQ QLAKRFCQYV VKQSELKGTF KGHKCSPCDT NWRYYGDSCY GFFRHNLTWE ESKQYCTDMN ATLLKIDNRN IVEYIKARTH LIRWVGLSRQ KSNEVWKWED GSVISENMFE FLEDGKGNMN CAYFHNGKMH PTFCENKHYL MCERKAGMTK VDQLP; Positive Samples: HepG2, HL-60, Mouse thymus, Mouse lung; Cellular Location: Membrane, Single-pass type II membrane protein.

Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CLEC1B
Gene Accession No. Q9JL99, Q9P126
Gene Alias 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384
Gene Symbols CLEC1B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-196 of human CLEC1B (NP_0010929011).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51266, 56760
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.