Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518938
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518938 Supplier Invitrogen™ Supplier No. PA518938
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CLEC1B
Gene Accession No. Q9P126
Gene Alias 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384
Gene Symbols CLEC1B
Host Species Goat
Immunogen Synthetic peptide sequence (KGHKCSPCDTNWRY) corresponding to the internal amino acids of CLEC1B (aa 95-109).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51266
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.