Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC1B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS2543R

Description
Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.Specifications
CLEC1B | |
Polyclonal | |
Unconjugated | |
CLEC1B | |
1810061I13Rik; BOS_5909; Clec1b; Clec2; CLEC-2; CLEC2B; CLEC9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member B; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; C-type lectin-like receptor 2; C-type lectin-like receptor-2; LOC100346882; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384 | |
Rabbit | |
Protein A | |
RUO | |
51266 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9P126 | |
CLEC1B | |
KLH conjugated synthetic peptide derived from human CLEC1B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction