Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC1B Monoclonal Antibody (AYP1), Biotin
GREENER_CHOICE

Catalog No. PIMA528529
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528529 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528529 Supplier Invitrogen™ Supplier No. MA528529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC1B Monoclonal Antibody for Flow, ICC/IF, IP

Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC1B
Applications Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone AYP1
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide; pH 7.4
Gene CLEC1B
Gene Accession No. Q9P126
Gene Alias 1810061I13Rik; Clec1b; CLEC2; CLEC-2; CLEC2B; CLEC9A; c-type lectin 1 member B; C-type lectin domain containing 9A; C-type lectin domain family 1 member B; C-type lectin domain family 1, member b; C-type lectin domain family 9 member A; C-type lectin domain family 1 member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; PRO1384; QDED721; RGD1563517; UNQ721/PRO1384
Gene Symbols CLEC1B
Host Species Mouse
Immunogen A recombinant extracellular fragment of human CLEC-2 (aa 68-229 extracellular domain).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.