Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLEC2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111692
Description
Immunogen sequence: IATWSKHAKP VACSGDWLGV RDKCFY.
CLEC2A (C-type lectin domain family 2 member A), also known as PILAR (proliferation-induced lymphocyte-associated receptor) is involved in modulating T-cell expansion. It is a 32 kDa single-pass type II membrane protein that contains one C-type lectin domain in its extracellular region and belongs to the CTL/CTLD superfamily. CLEC2A is mainly expressed in skin and highly expressed in CD8(+), B lymphocytes and naive CD4(+) T cells. Manipulation of CLEC2A signaling may be important for treatment of autoimmune diseases and cancer.
Specifications
| CLEC2A | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UVW9 | |
| CLEC2A | |
| Recombinant Protein corresonding to Human CLEC2A. Recombinant protein control fragment (Product #RP-107877). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| CLEC2A | |
| CLEC2A; C-type lectin domain family 2 member A; C-type lectin domain family 2, member A; INPE5792; KACL; Keratinocyte-associated C-type lectin; PILAR; Proliferation-induced lymphocyte-associated receptor; UNQ579; UNQ5792; UNQ5792/PRO19597 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387836 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction