Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116747 Supplier Invitrogen™ Supplier No. PA5116747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat spleen Immunogen sequence: IGFQNKCYYF SKEEGDWNSS KYNCSTQHAD LTIIDNIEEM NFLRRYKCSS DHWIGLKMAK NRTGQWVDGA TFTKSFGMRG S.

CLEC2B encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell activation antigen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLEC2B
Gene Accession No. Q92478
Gene Alias Activation-induced C-type lectin; AICL; CLEC2B; CLECSF2; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 2 (activation-induced); C-type lectin domain family 2 member B; C-type lectin domain family 2, member B; C-type lectin superfamily member 2; HP10085; IFN-alpha2b-inducing related protein 1; IFN-alpha-2b-inducing-related protein 1; IFNRG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human CLEC2B (NP_0051182).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9976
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.