Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560911 Supplier Invitrogen™ Supplier No. PA560911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEDWLLYGRK CYFFSEEPRD WNTGRQYCHT HEAVLAVIQS QKELEFMFKF TRREPWIGLR RVGDEFHWVN GDPFDPDTFT IAGPGECVFV EP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC2L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLEC2L
Gene Accession No. P0C7M8
Gene Alias AI414054; AW122724; Clec2l; C-type lectin domain family 2 member L; C-type lectin domain family 2, member L; C-type lectin domain family, member L; Gm1064
Gene Symbols CLEC2L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLEC2L. Recombinant protein control fragment (Product #RP-100573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.