Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570780 Supplier Invitrogen™ Supplier No. PA570780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 79%.

CLEC3A gene ontology annotations related to this gene include carbohydrate binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC3A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CLEC3A
Gene Accession No. O75596
Gene Alias 1110019O10Rik; Cartilage-derived C-type lectin; clec3a; CLECSF1; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 1 (cartilage-derived); C-type calcium dependent, carbohydrate-recognition domain lectin, superfamily member 1, cartilage-derived; C-type lectin domain family 3 member A; C-type lectin domain family 3, member a; C-type lectin homolog precursor; C-type lectin superfamily member 1; domain) lectin, superfamily member 1(cartilage-derived); Gm796; UNQ700/PRO1345
Gene Symbols CLEC3A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human CLEC3A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.