Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLEC3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597595
Description
CLEC3A Polyclonal Antibody for Western Blot, IHC (P), ELISA
CLEC3A gene ontology annotations related to this gene include carbohydrate binding.
Specifications
CLEC3A | |
Polyclonal | |
Unconjugated | |
CLEC3A | |
1110019O10Rik; Cartilage-derived C-type lectin; clec3a; CLECSF1; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 1 (cartilage-derived); C-type calcium dependent, carbohydrate-recognition domain lectin, superfamily member 1, cartilage-derived; C-type lectin domain family 3 member A; C-type lectin domain family 3, member a; C-type lectin homolog precursor; C-type lectin superfamily member 1; domain) lectin, superfamily member 1(cartilage-derived); Gm796; UNQ700/PRO1345 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10143, 403395 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O75596, Q9EPW4 | |
CLEC3A | |
Recombinant Human C-type lectin domain family 3 member A protein (23-197AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction