Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC3B/Tetranectin Antibody (008), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290148R
Description
CLEC3B/Tetranectin Monoclonal specifically detects CLEC3B/Tetranectin in Human samples. It is validated for ELISA.Specifications
CLEC3B/Tetranectin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse CLEC14A | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
8 | |
ELISA | |
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CLEC3B/Tetranectin (AAX37102.1; Met1-Val202). | |
0.1 mL | |
Cancer | |
7123 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction