Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC3B/Tetranectin Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298345APC
Description
CLEC3B/Tetranectin Polyclonal antibody specifically detects CLEC3B/Tetranectin in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
CLEC3B/Tetranectin | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin | |
Produced in rabbits immunized with purified, recombinant Mouse CLEC3B/Tetranectin (Uniprot#: P43025; Met1-Val202) | |
0.1 mL | |
Cancer | |
7123 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction