Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC4A Monoclonal Antibody (1E3A4)
GREENER_CHOICE

Catalog No. PIMA529154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529154 Supplier Invitrogen™ Supplier No. MA529154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DCIR/CLEC4A.

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in inflammatory and immune response. Multiple transcript variants encoding distinct isoforms have been identified for this gene. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4A
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 1E3A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CLEC4A
Gene Accession No. Q9UMR7
Gene Alias CD367; CLEC4A; Clec4a2; Clecsf6; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 6; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 6; C-type lectin DDB27; C-type lectin domain family 4 member A; C-type lectin domain family 4, member A; C-type lectin domain family 4, member a2; C-type lectin superfamily member 6; DCIR; Dcir1; Dcir2; DDB27; Dendritic cell immunoreceptor; dendritic cell inhibitory receptor 1; dendritic cell inhibitory receptor 2; HDCGC13P; immunoreceptor; lectin-like immunoreceptor; LLIR
Gene Symbols CLEC4A
Host Species Mouse
Immunogen Recombinant Human CLEC4A protein (Gln70-Leu237).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.