Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC4C Monoclonal Antibody (3F9B1)
GREENER_CHOICE

Catalog No. FERMA548812
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548812 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548812 Supplier Invitrogen™ Supplier No. MA548812
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4C Monoclonal Antibody for Western Blot, Flow, ELISA

This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4C
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3F9B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CLEC4C
Gene Accession No. Q8WTT0
Gene Alias BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150
Gene Symbols CLEC4C
Host Species Mouse
Immunogen Purified recombinant fragment of human CD303 (AA: extra 45-213) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.